missing translation for 'onlineSavingsMsg'
Learn More

3-Iodotoluene, 99%

CAS: 625-95-6 | C7H7I | 218.037 g/mol

Supplier:  Thermo Scientific Chemicals A1099922

 View more versions of this product

Catalog No. AAA1099922


Only null left
Explore available promotions

Description

Description

3-Iodotoluene was used in the preparartion of indene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodotoluene was used in the preparartion of indene derivatives.

Solubility
Not miscible in water. Soluble in benzene, alcohol and ether.

Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

3-Iodotoluene
-27°C
80°C to 82°C (10 mmHg)
C7H7I
MFCD00001050
1903634
3-iodotoluene, m-iodotoluene, benzene, 1-iodo-3-methyl, m-tolyl iodide, m-methyliodobenzene, toluene, m-iodo, 3-methylphenyl iodide, unii-2h574ir35g, 1-iodo-3-methyl-benzene, m-iodo-toluen
VLCPISYURGTGLP-UHFFFAOYSA-N
1-iodo-3-methylbenzene
12268
99%
625-95-6
1.698
82°C (179°F)
1.604
100 g
Light sensitive
Not miscible in water. Soluble in benzene,alcohol and ether.
CC1=CC(=CC=C1)I
218.037
218.04

Chemical Identifiers

625-95-6
218.037
VLCPISYURGTGLP-UHFFFAOYSA-N
12268
CC1=CC(=CC=C1)I
C7H7I
MFCD00001050
3-iodotoluene, m-iodotoluene, benzene, 1-iodo-3-methyl, m-tolyl iodide, m-methyliodobenzene, toluene, m-iodo, 3-methylphenyl iodide, unii-2h574ir35g, 1-iodo-3-methyl-benzene, m-iodo-toluen
1-iodo-3-methylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 210-918-5

RTECSNumber : DA3440000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only