missing translation for 'onlineSavingsMsg'
Learn More

3-Iodotoluene, 99%

Catalog No. AAA1099918
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1099918 50 g
AAA1099922 100 g
AAA1099930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1099918 Supplier Thermo Scientific Chemicals Supplier No. A1099918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 625-95-6 | C7H7I | 218.037 g/mol

3-Iodotoluene was used in the preparartion of indene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodotoluene was used in the preparartion of indene derivatives.

Solubility
Not miscible in water. Soluble in benzene, alcohol and ether.

Notes
Light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 625-95-6
Molecular Formula C7H7I
Molecular Weight (g/mol) 218.037
MDL Number MFCD00001050
InChI Key VLCPISYURGTGLP-UHFFFAOYSA-N
Synonym 3-iodotoluene, m-iodotoluene, benzene, 1-iodo-3-methyl, m-tolyl iodide, m-methyliodobenzene, toluene, m-iodo, 3-methylphenyl iodide, unii-2h574ir35g, 1-iodo-3-methyl-benzene, m-iodo-toluen
PubChem CID 12268
IUPAC Name 1-iodo-3-methylbenzene
SMILES CC1=CC(=CC=C1)I

Specifications

Melting Point -27°C
Density 1.698
Boiling Point 80°C to 82°C (10 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.604
Quantity 50 g
Beilstein 1903634
Sensitivity Light sensitive
Solubility Information Not miscible in water. Soluble in benzene,alcohol and ether.
Formula Weight 218.04
Percent Purity 99%
Chemical Name or Material 3-Iodotoluene
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 210-918-5
RTECSNumber DA3440000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only