missing translation for 'onlineSavingsMsg'
Learn More

3-iodotetrahydrofuran, 97%, Thermo Scientific™

Catalog No. CC76011DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC76011DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC76011DA Supplier Thermo Scientific Chemicals Supplier No. CC76011DA

Chemical Identifiers

CAS 121138-01-0
Molecular Formula C4H7IO
Molecular Weight (g/mol) 198.00
MDL Number MFCD09878873
InChI Key BKIQORJIKOPRCG-UHFFFAOYNA-N
Synonym 3-iodotetrahydrofuran, 3-iodo-tetrahydro-furan, furan, tetrahydro-3-iodo, 3-iodanyloxolane, iodotetrahydrofuran
PubChem CID 22139264
IUPAC Name 3-iodooxolane
SMILES IC1CCOC1

Specifications

Color Colorless
CAS Min % 96.5
CAS Max % 100.0
Quantity 1 g
Packaging Amber Glass Bottle
Physical Form Liquid
Assay Percent Range 97%
Boiling Point 110°C
Formula Weight 198
Percent Purity 97%
Chemical Name or Material 3-Iodotetrahydrofuran
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.