missing translation for 'onlineSavingsMsg'
Learn More

3-Iodopyridine 98.0+%, TCI America™

Catalog No. p-7131490
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
I05685G 5 g
I056825G 25 g
2 options

Catalog No. I05685G

Supplier: TCI America I05685G

Only null left

Chemical Identifiers

CAS 1120-90-7
Molecular Formula C5H4IN
Molecular Weight (g/mol) 204.998
MDL Number MFCD00023553
InChI Key XDELKSRGBLWMBA-UHFFFAOYSA-N
Synonym 3-iodo pyridine, 3-iodo-pyridine, pyridine, 3-iodo, 3-iodpyridin, 3-iodopyridine, pubchem6634, acmc-1bvap, ksc490s0t
PubChem CID 70714
IUPAC Name 3-iodopyridine
SMILES C1=CC(=CN=C1)I

Specifications

Melting Point 55°C
Boiling Point 92°C
Quantity 5 g
Formula Weight 205.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodopyridine