missing translation for 'onlineSavingsMsg'
Learn More

3-Iodopyridine, 99%

Catalog No. p-7026914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2322506 5 g
AAB2322514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2322506 Supplier Thermo Scientific Chemicals Supplier No. B2322506

CAS: 1120-90-7 | C5H4IN | 204.998 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1120-90-7
Molecular Formula C5H4IN
Molecular Weight (g/mol) 204.998
MDL Number MFCD00023553
InChI Key XDELKSRGBLWMBA-UHFFFAOYSA-N
Synonym 3-iodo pyridine, 3-iodo-pyridine, pyridine, 3-iodo, 3-iodpyridin, 3-iodopyridine, pubchem6634, acmc-1bvap, ksc490s0t
PubChem CID 70714
IUPAC Name 3-iodopyridine
SMILES C1=CC(=CN=C1)I

Specifications

Melting Point 53°C to 56°C
Boiling Point 90°C to 92°C (14 mmHg)
Flash Point 106°C (222°F)
Quantity 5 g
Beilstein 106059
Sensitivity Light sensitive
Formula Weight 205
Percent Purity 99%
Chemical Name or Material 3-Iodopyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-322-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only