missing translation for 'onlineSavingsMsg'
Learn More

3-Iodopropionic acid, 99%

Catalog No. AAL0265506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0265506 5 g
AAL0265514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0265506 Supplier Thermo Scientific Chemicals Supplier No. L0265506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 141-76-4 | C3H5IO2 | 199.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141-76-4
Molecular Formula C3H5IO2
Molecular Weight (g/mol) 199.98
MDL Number MFCD00002765
InChI Key KMRNTNDWADEIIX-UHFFFAOYSA-N
Synonym 3-iodopropionic acid, propanoic acid, 3-iodo, beta-iodopropionic acid, propionic acid, 3-iodo, iodopropanoic acid, 3-iodo-propionic acid, .beta.-iodopropionic acid, iodo-propanoic acid, b-iodopropionic acid, 3-iodo-propanoic acid
PubChem CID 8856
IUPAC Name 3-iodopropanoic acid
SMILES OC(=O)CCI

Specifications

Melting Point 80°C to 83°C
Quantity 5 g
UN Number UN3261
Beilstein 1699514
Sensitivity Light sensitive
Formula Weight 199.98
Percent Purity 99%
Chemical Name or Material 3-Iodopropionic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 205-499-0
RTECSNumber UF4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only