missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophthalic acid, ≥95%, Thermo Scientific™

Catalog No. BTB12976EA
Change view
Click to view available options
Quantity:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB12976EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB12976EA Supplier Thermo Scientific Chemicals Supplier No. BTB12976EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6937-34-4
Molecular Formula C8H5IO4
Molecular Weight (g/mol) 292.03
MDL Number MFCD00275568
InChI Key HNPVERUJGFNNRV-UHFFFAOYSA-N
Synonym 3-iodobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylicacid, 3-iodo, maybridge1_008994, 3-iodo-phthalic acid, 1,2-benzenedicarboxylic acid, 3-iodo
PubChem CID 238231
IUPAC Name 3-iodophthalic acid
SMILES OC(=O)C1=CC=CC(I)=C1C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula IC6H3(CO2H)2
Quantity 10g
Formula Weight 292.02
Percent Purity 97%
Chemical Name or Material 3-Iodophthalic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.