accessibility menu, dialog, popup

UserName

HOOIOHO

CAS RN 6937-34-4

IUPAC Name: 3-iodobenzene-1,2-dicarboxylic acid
Synonyms: 3-iodobenzene-1,2-dicarboxylic acid
Molecular Weight (g/mol): 292.03
Molecular Formula: C8H5IO4
InChi Key: HNPVERUJGFNNRV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(I)=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)