missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7131583
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07601G 1 g
I07605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07601G Supplier TCI America Supplier No. I07601G

Chemical Identifiers

CAS 1878-69-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.05
MDL Number MFCD00046548
InChI Key MRSWWBAFGGGWRH-UHFFFAOYSA-N
Synonym 3-iodophenylacetic acid, 2-3-iodophenyl acetic acid, 3-iodophenyl acetic acid, m-iodophenylacetic acid, m-iodobenzeneacetic acid, benzeneacetic acid, 3-iodo, pubchem4195, 3-iodophenylaceticacid, acmc-1cfam, 3-iodobenzeneacetic acid
PubChem CID 3870220
IUPAC Name 2-(3-iodophenyl)acetic acid
SMILES OC(=O)CC1=CC(I)=CC=C1

Specifications

Melting Point 131°C
Quantity 1 g
Formula Weight 262.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodophenylacetic Acid