missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophenylacetic acid, 98%

Catalog No. AAL1511704
Change view
Click to view available options
Quantity:
500 mg
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1511704 2 g
AAL15117ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1511704 Supplier Thermo Scientific Chemicals Supplier No. L1511704
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1878-69-9 | C8H7IO2 | 262.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1878-69-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.05
MDL Number MFCD00046548
InChI Key MRSWWBAFGGGWRH-UHFFFAOYSA-N
Synonym 3-iodophenylacetic acid, 2-3-iodophenyl acetic acid, 3-iodophenyl acetic acid, m-iodophenylacetic acid, m-iodobenzeneacetic acid, benzeneacetic acid, 3-iodo, pubchem4195, 3-iodophenylaceticacid, acmc-1cfam, 3-iodobenzeneacetic acid
PubChem CID 3870220
IUPAC Name 2-(3-iodophenyl)acetic acid
SMILES OC(=O)CC1=CC(I)=CC=C1

Specifications

Melting Point 128°C to 132°C
Quantity 2 g
Beilstein 2501867
Sensitivity Light sensitive
Formula Weight 262.05
Percent Purity 98%
Chemical Name or Material 3-Iodophenylacetic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only