missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. I09855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09855G 5 g
I09851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09855G Supplier TCI America Supplier No. I09855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3125-73-3
Molecular Formula C7H4INS
Molecular Weight (g/mol) 261.08
MDL Number MFCD00041074
InChI Key LKCSXVAACHUTGC-UHFFFAOYSA-N
Synonym 3-iodophenyl isothiocyanate, 3-iodophenylisothiocyanate, benzene, 1-iodo-3-isothiocyanato, m-iodophenylisothiocyanate, 3-ipi, isothiocyanic acid, m-iodophenyl ester, acmc-1cmge, benzene,1-iodo-3-isothiocyanato, 1-iodanyl-3-isothiocyanato-benzene
PubChem CID 151083
IUPAC Name 1-iodo-3-isothiocyanatobenzene
SMILES IC1=CC=CC(=C1)N=C=S

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 5 g
UN Number 2923
Formula Weight 261.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Iodophenyl Isothiocyanate