missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophenol 98.0+%, TCI America™

Catalog No. p-7131440
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I049510G 10 g
I049525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I049510G Supplier TCI America Supplier No. I049510G
Only null left

Chemical Identifiers

CAS 626-02-8
Molecular Formula C6H5IO
Molecular Weight (g/mol) 220.009
MDL Number MFCD00002261
InChI Key FXTKWBZFNQHAAO-UHFFFAOYSA-N
Synonym m-iodophenol, phenol, 3-iodo, m-hydroxyiodobenzene, 3-jodphenol, phenol, m-iodo, 3-jodphenol german, m-jodphenol, 3-iodo-phenol, 5-iodophenol, pubchem3968
PubChem CID 12272
ChEBI CHEBI:33439
IUPAC Name 3-iodophenol
SMILES C1=CC(=CC(=C1)I)O

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 190°C
Quantity 10 g
Formula Weight 220.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodophenol
RTECSNumber SL5490000
TSCA Yes