missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzyl Alcohol 98.0+%, TCI America™

Catalog No. p-7131636
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08665G 5 g
I086625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08665G Supplier TCI America Supplier No. I08665G
Only null left

Chemical Identifiers

CAS 57455-06-8
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD00004635
InChI Key QGCCNWSXJHGUNL-UHFFFAOYSA-N
Synonym 3-iodophenyl methanol, 3-iodobenzyl alcohol, m-iodobenzyl alcohol, benzenemethanol, 3-iodo, benzyl alcohol, m-iodo, 3-iodo-benzyl alcohol, 3-iodobenzylalcohol, 3-iodo-phenyl-methanol, m-iodo benzylalcohol, 3-iodophenyl methan-1-ol
PubChem CID 42300
IUPAC Name (3-iodophenyl)methanol
SMILES C1=CC(=CC(=C1)I)CO

Specifications

Color Yellow
Boiling Point 105°C
Quantity 5 g
Formula Weight 234.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Iodobenzyl Alcohol
RTECSNumber DO8318000
TSCA No