missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzoic acid, 98%

Catalog No. p-149338
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160480100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC160480100 Supplier Thermo Scientific Chemicals Supplier No. 160480100

CAS: 618-51-9 | C7H5IO2 | 248.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 618-51-9
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.01
MDL Number MFCD00002496
InChI Key KVBWBCRPWVKFQT-UHFFFAOYSA-N
Synonym m-iodobenzoic acid, benzoic acid, 3-iodo, 3-iodo-benzoic acid, benzoic acid, m-iodo, 3-iodobenzoicacid, 3-carboxyiodobenzene, 3-iodo benzoic acid, pubchem3925, acmc-209mwv, meta-carboxy iodo-benzene
PubChem CID 12060
IUPAC Name 3-iodobenzoic acid
SMILES C1=CC(=CC(=C1)I)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 185°C to 189°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula IC6H4CO2H
Quantity 10 g
Beilstein 09, 365
Solubility Information Solubility in water: slightly soluble
Formula Weight 248.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Iodobenzoic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
EINECSNumber 210-555-2
TSCA TSCA

RUO – Research Use Only