missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzoic acid, 98+%

Catalog No. AAA1151509
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1151509 10 g
AAA1151518 50 g
AAA1151530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1151509 Supplier Thermo Scientific Chemicals Supplier No. A1151509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 618-51-9 | C7H5IO2 | 248.019 g/mol

3-Iodobenzoic acid is used in solid phase synthesis of γ-turn mimetic library. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodobenzoic acid is used in solid phase synthesis of γ-turn mimetic library. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in Chloroform and Methanol. Insoluble in water.

Notes
Light Sensitive. Store in the dark. Incompatible materials are bases and oxidizing agents.

Chemical Identifiers

CAS 618-51-9
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.019
MDL Number MFCD00002496
InChI Key KVBWBCRPWVKFQT-UHFFFAOYSA-N
Synonym m-iodobenzoic acid, benzoic acid, 3-iodo, 3-iodo-benzoic acid, benzoic acid, m-iodo, 3-iodobenzoicacid, 3-carboxyiodobenzene, 3-iodo benzoic acid, pubchem3925, acmc-209mwv, meta-carboxy iodo-benzene
PubChem CID 12060
IUPAC Name 3-iodobenzoic acid
SMILES C1=CC(=CC(=C1)I)C(=O)O

Specifications

Melting Point 186°C to 188°C
Quantity 10 g
Beilstein 971088
Sensitivity Light sensitive
Solubility Information Soluble in Chloroform and Methanol. Insoluble in water.
Formula Weight 248.02
Percent Purity ≥98%
Chemical Name or Material 3-Iodobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-555-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only