missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzeneboronic acid pinacol ester, 97%

Catalog No. AAH2811503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2811503 1 g
AAH2811506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2811503 Supplier Thermo Scientific Chemicals Supplier No. H2811503

CAS: 408492-28-4 | C12H16BIO2 | 329.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 408492-28-4
Molecular Formula C12H16BIO2
Molecular Weight (g/mol) 329.97
MDL Number MFCD09953498
InChI Key HSHFNMSHDHAHDN-UHFFFAOYSA-N
Synonym 2-3-iodophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-iodophenylboronic acid pinacol ester, amtb088, 3-iodophenylboronic acid pinacolester, 1,3,2-dioxaborolane,2-3-iodophenyl-4,4,5,5-tetramethyl, 2-3-iodanylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-iodo-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene
PubChem CID 11313483
IUPAC Name 2-(3-iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC(I)=CC=C1

Specifications

Melting Point 70°C to 72°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 329.97
Percent Purity 97%
Chemical Name or Material 3-Iodobenzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only