Learn More
3-Iodobenzeneboronic acid, 97%
CAS: 221037-98-5 | C6H6BIO2 | 247.826 g/mol
$51.19 - $171.07
Chemical Identifiers
| CAS | 221037-98-5 |
|---|---|
| Molecular Formula | C6H6BIO2 |
| Molecular Weight (g/mol) | 247.826 |
| MDL Number | MFCD01319015 |
| InChI Key | REEUXWXIMNEIIN-UHFFFAOYSA-N |
| Synonym | 3-iodophenyl boronic acid, boronic acid, 3-iodophenyl, 3-iodobenzeneboronic acid, pentanedioic acid 1-1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl ester, pubchem6224, acmc-1ckkp, 3-iodophenylboranic acid, amtb078, 3-iodanylphenyl boronic acid, boronic acid,b-3-iodophenyl |
| PubChem CID | 2734362 |
| IUPAC Name | (3-iodophenyl)boronic acid |
| SMILES | B(C1=CC(=CC=C1)I)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5111203
|
Thermo Scientific Chemicals
H5111203 |
1 g |
Each for $51.19
|
|
|||||
|
AAH5111206
|
Thermo Scientific Chemicals
H5111206 |
5 g |
Each for $171.07
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 221037-98-5 | |
| 247.826 | |
| REEUXWXIMNEIIN-UHFFFAOYSA-N | |
| 2734362 | |
| B(C1=CC(=CC=C1)I)(O)O |
| C6H6BIO2 | |
| MFCD01319015 | |
| 3-iodophenyl boronic acid, boronic acid, 3-iodophenyl, 3-iodobenzeneboronic acid, pentanedioic acid 1-1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl ester, pubchem6224, acmc-1ckkp, 3-iodophenylboranic acid, amtb078, 3-iodanylphenyl boronic acid, boronic acid,b-3-iodophenyl | |
| (3-iodophenyl)boronic acid |
Specifications
| 221037-98-5 | |
| MFCD01319015 | |
| Light sensitive | |
| REEUXWXIMNEIIN-UHFFFAOYSA-N | |
| (3-iodophenyl)boronic acid | |
| 2734362 | |
| 97% |
| C6H6BIO2 | |
| 1 g | |
| 3-iodophenyl boronic acid, boronic acid, 3-iodophenyl, 3-iodobenzeneboronic acid, pentanedioic acid 1-1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl ester, pubchem6224, acmc-1ckkp, 3-iodophenylboranic acid, amtb078, 3-iodanylphenyl boronic acid, boronic acid,b-3-iodophenyl | |
| B(C1=CC(=CC=C1)I)(O)O | |
| 247.826 | |
| 247.83 | |
| 3-Iodobenzeneboronic acid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only