missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzeneboronic acid, 97%

Catalog No. AAH5111203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5111203 1 g
AAH5111206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5111203 Supplier Thermo Scientific Chemicals Supplier No. H5111203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 221037-98-5 | C6H6BIO2 | 247.826 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 221037-98-5
Molecular Formula C6H6BIO2
Molecular Weight (g/mol) 247.826
MDL Number MFCD01319015
InChI Key REEUXWXIMNEIIN-UHFFFAOYSA-N
Synonym 3-iodophenyl boronic acid, boronic acid, 3-iodophenyl, 3-iodobenzeneboronic acid, pentanedioic acid 1-1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl ester, pubchem6224, acmc-1ckkp, 3-iodophenylboranic acid, amtb078, 3-iodanylphenyl boronic acid, boronic acid,b-3-iodophenyl
PubChem CID 2734362
IUPAC Name (3-iodophenyl)boronic acid
SMILES B(C1=CC(=CC=C1)I)(O)O

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 247.83
Percent Purity 97%
Chemical Name or Material 3-Iodobenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only