missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzaldehyde, 99%

Catalog No. AC392070050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392070050 5 g Glass bottle
AC392070010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC392070050 Supplier Thermo Scientific Chemicals Supplier No. 392070050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 696-41-3 | C7H5IO | 232.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 696-41-3
Molecular Formula C7H5IO
Molecular Weight (g/mol) 232.02
MDL Number MFCD00039573
InChI Key RZODAQZAFOBFLS-UHFFFAOYSA-N
Synonym 3-iodo-benzaldehyde, 3-iodbenzaldehyd, benzaldehyde, 3-iodo, 3-iodo benzaldehyde, 3-iodanylbenzaldehyde, pubchem3917, acmc-1atsd, 1-formyl-3-iodobenzene, m-iodobenzaldehyde, 3-iodobenzaldehyde
PubChem CID 252610
IUPAC Name 3-iodobenzaldehyde
SMILES C1=CC(=CC(=C1)I)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 57.0°C to 60.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4CHO
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 232.02
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material 3-Iodobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only