missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodoanisole 98.0+%, TCI America™
$44.28 - $144.39
Chemical Identifiers
| CAS | 766-85-8 |
|---|---|
| Molecular Formula | C7H7IO |
| Molecular Weight (g/mol) | 234.036 |
| MDL Number | MFCD00001048 |
| InChI Key | RSHBAGGASAJQCH-UHFFFAOYSA-N |
| Synonym | 3-iodoanisole, m-iodoanisole, benzene, 1-iodo-3-methoxy, anisole, m-iodo, m-methoxyiodobenzene, 3-methoxyphenyl iodide, 3-methoxyiodobenzene, 1-iodo-3-methoxy-benzene, m-iodoanisol, m-iodo-anisole |
| PubChem CID | 69839 |
| IUPAC Name | 1-iodo-3-methoxybenzene |
| SMILES | COC1=CC(=CC=C1)I |
Chemical Identifiers
| 766-85-8 | |
| 234.036 | |
| RSHBAGGASAJQCH-UHFFFAOYSA-N | |
| 69839 | |
| COC1=CC(=CC=C1)I |
| C7H7IO | |
| MFCD00001048 | |
| 3-iodoanisole, m-iodoanisole, benzene, 1-iodo-3-methoxy, anisole, m-iodo, m-methoxyiodobenzene, 3-methoxyphenyl iodide, 3-methoxyiodobenzene, 1-iodo-3-methoxy-benzene, m-iodoanisol, m-iodo-anisole | |
| 1-iodo-3-methoxybenzene |
Specifications
| 766-85-8 | |
| 245°C | |
| MFCD00001048 | |
| 3-iodoanisole, m-iodoanisole, benzene, 1-iodo-3-methoxy, anisole, m-iodo, m-methoxyiodobenzene, 3-methoxyphenyl iodide, 3-methoxyiodobenzene, 1-iodo-3-methoxy-benzene, m-iodoanisol, m-iodo-anisole | |
| COC1=CC(=CC=C1)I | |
| 234.036 | |
| 234.04 | |
| Liquid |
| Yellow | |
| C7H7IO | |
| 5 g | |
| RSHBAGGASAJQCH-UHFFFAOYSA-N | |
| 1-iodo-3-methoxybenzene | |
| 69839 | |
| ≥98.0% (GC) | |
| 3-Iodoanisole |
Safety and Handling
TSCA : Yes