Learn More
3-Iodoanisole, 98%
CAS: 766-85-8 | C7H7IO | 234.036 g/mol
$54.73 - $494.29
Chemical Identifiers
| CAS | 766-85-8 |
|---|---|
| Molecular Formula | C7H7IO |
| Molecular Weight (g/mol) | 234.036 |
| MDL Number | MFCD00001048 |
| InChI Key | RSHBAGGASAJQCH-UHFFFAOYSA-N |
| Synonym | 3-iodoanisole, m-iodoanisole, benzene, 1-iodo-3-methoxy, anisole, m-iodo, m-methoxyiodobenzene, 3-methoxyphenyl iodide, 3-methoxyiodobenzene, 1-iodo-3-methoxy-benzene, m-iodoanisol, m-iodo-anisole |
| PubChem CID | 69839 |
| IUPAC Name | 1-iodo-3-methoxybenzene |
| SMILES | COC1=CC(=CC=C1)I |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1420806
|
Thermo Scientific Chemicals
A1420806 |
5 g |
Each for $54.73
|
|
|||||
|
AAA1420814
|
Thermo Scientific Chemicals
A1420814 |
25 g |
Each for $171.07
|
|
|||||
|
AAA1420822
|
Thermo Scientific Chemicals
A1420822 |
100 g |
Each for $494.29
|
|
|||||
Description
3-Iodoanisole is used to produce 3,3'-Dimethoxy-biphenyl. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Iodoanisole is used to produce 3,3′-Dimethoxy-biphenyl. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in chloroform, alcohol, ether. Insoluble in water.
Notes
Light Sensitive. Incompatible with light and oxidizing agents. Store in the dark.
Chemical Identifiers
| 766-85-8 | |
| 234.036 | |
| RSHBAGGASAJQCH-UHFFFAOYSA-N | |
| 69839 | |
| COC1=CC(=CC=C1)I |
| C7H7IO | |
| MFCD00001048 | |
| 3-iodoanisole, m-iodoanisole, benzene, 1-iodo-3-methoxy, anisole, m-iodo, m-methoxyiodobenzene, 3-methoxyphenyl iodide, 3-methoxyiodobenzene, 1-iodo-3-methoxy-benzene, m-iodoanisol, m-iodo-anisole | |
| 1-iodo-3-methoxybenzene |
Specifications
| 766-85-8 | |
| 243°C to 244°C | |
| C7H7IO | |
| MFCD00001048 | |
| 1858896 | |
| 3-iodoanisole, m-iodoanisole, benzene, 1-iodo-3-methoxy, anisole, m-iodo, m-methoxyiodobenzene, 3-methoxyphenyl iodide, 3-methoxyiodobenzene, 1-iodo-3-methoxy-benzene, m-iodoanisol, m-iodo-anisole | |
| RSHBAGGASAJQCH-UHFFFAOYSA-N | |
| 1-iodo-3-methoxybenzene | |
| 69839 | |
| 98% |
| 1.78 | |
| >110°C (230°F) | |
| 1.613 | |
| 5 g | |
| Light sensitive | |
| Soluble in chloroform,alcohol,ether. Insoluble in water. | |
| COC1=CC(=CC=C1)I | |
| 234.036 | |
| 234.04 | |
| 3-Iodoanisole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H320-H335
EINECSNumber : 212-173-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only