missing translation for 'onlineSavingsMsg'
Learn More

3-Iodoanisole, 98%

Catalog No. AAA1420822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1420822 100 g
AAA1420806 5 g
AAA1420814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1420822 Supplier Thermo Scientific Chemicals Supplier No. A1420822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 766-85-8 | C7H7IO | 234.036 g/mol

3-Iodoanisole is used to produce 3,3'-Dimethoxy-biphenyl. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodoanisole is used to produce 3,3′-Dimethoxy-biphenyl. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in chloroform, alcohol, ether. Insoluble in water.

Notes
Light Sensitive. Incompatible with light and oxidizing agents. Store in the dark.

Chemical Identifiers

CAS 766-85-8
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD00001048
InChI Key RSHBAGGASAJQCH-UHFFFAOYSA-N
Synonym 3-iodoanisole, m-iodoanisole, benzene, 1-iodo-3-methoxy, anisole, m-iodo, m-methoxyiodobenzene, 3-methoxyphenyl iodide, 3-methoxyiodobenzene, 1-iodo-3-methoxy-benzene, m-iodoanisol, m-iodo-anisole
PubChem CID 69839
IUPAC Name 1-iodo-3-methoxybenzene
SMILES COC1=CC(=CC=C1)I

Specifications

Density 1.78
Boiling Point 243°C to 244°C
Flash Point >110°C (230°F)
Refractive Index 1.613
Quantity 100 g
Beilstein 1858896
Sensitivity Light sensitive
Solubility Information Soluble in chloroform,alcohol,ether. Insoluble in water.
Formula Weight 234.04
Percent Purity 98%
Chemical Name or Material 3-Iodoanisole
Show More Show Less
Hazard Category H315-H320-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-173-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only