missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-o-xylene, 97%

Catalog No. AAA1177414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1177414 25 g
AAA1177406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1177414 Supplier Thermo Scientific Chemicals Supplier No. A1177414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 31599-60-7 | C8H9I | 232.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31599-60-7
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00041437
InChI Key DANMWBNOPFBJSZ-UHFFFAOYSA-N
Synonym 3-iodo-o-xylene, 1,2-dimethyl-3-iodobenzene, 2,3-dimethyliodobenzene, benzene, 1-iodo-2,3-dimethyl, benzene, iododimethyl, 2,3-dimethyl-1-iodobenzene, jodxylol, 3-iodo-ortho-xylene, pubchem3865, acmc-1ajf2
PubChem CID 141645
IUPAC Name 1-iodo-2,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)I)C

Specifications

Density 1.64
Quantity 25 g
Refractive Index 1.603
Beilstein 1929733
Boiling Point 98°C to 100°C (15 mmHg)
Formula Weight 232.07
Percent Purity 97%
Chemical Name or Material 3-Iodo-o-xylene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only