missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-L-tyrosine 98.0+%, TCI America™

Catalog No. p-7131197
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00751G 1 g
I00755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I00751G Supplier TCI America Supplier No. I00751G
Only null left

Chemical Identifiers

CAS 70-78-0
Molecular Formula C9H10INO3
Molecular Weight (g/mol) 307.09
MDL Number MFCD00002608
InChI Key UQTZMGFTRHFAAM-JLDDOWRYNA-N
Synonym 3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanine
PubChem CID 439744
ChEBI CHEBI:27847
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O

Specifications

Melting Point 199°C
Color White-Yellow
Quantity 1 g
Formula Weight 307.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodo-L-tyrosine
TSCA No
Recommended Storage Refrigerator