missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-4-nitroanisole, 97%

Catalog No. AC326320050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC326320050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC326320050 Supplier Thermo Scientific Chemicals Supplier No. 326320050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 214279-40-0 | C7H6INO3 | 279.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 214279-40-0
Molecular Formula C7H6INO3
Molecular Weight (g/mol) 279.03
MDL Number MFCD01320682
InChI Key NLTAHTYBZZSCIT-UHFFFAOYSA-N
Synonym 3-iodo-4-nitroanisole, 4-nitro-3-iodoanisole, benzene, 2-iodo-4-methoxy-1-nitro, 1-nitro-2-iodo-4-methoxybenzene, 3-iodo-4-nitrophenyl methyl ether, benzene,2-iodo-4-methoxy-1-nitro
PubChem CID 2733456
IUPAC Name 2-iodo-4-methoxy-1-nitrobenzene
SMILES COC1=CC=C(C(I)=C1)[N+]([O-])=O

Specifications

Melting Point 68.0°C to 73.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 279.03
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Iodo-4-nitroanisole
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only