missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-4-methylaniline, 98%

Catalog No. p-7081432
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1148506 5 g
AAA1148514 25 g
AAA1148522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1148506 Supplier Thermo Scientific Chemicals Supplier No. A1148506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35944-64-0 | C7H8IN | 233.05 g/mol

3-Iodo-4-methylaniline is used as fine chemicals, pharmaceutical intermediates. 2-[((3-iodo-4-methyl)phenylimino)methyl]-5-nitrothiophene was derived from 5-nitro-2-thiophene-carboxaldehyde and 3-iodo-4-methylaniline. 1-(3-iodo-4-methylphenyl)-3-tert-butyl-2H-aminopyrazole (5), prepared from 3-iodo-4-methylaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodo-4-methylaniline is used as fine chemicals, pharmaceutical intermediates. 2-[((3-iodo-4-methyl)phenylimino)methyl]-5-nitrothiophene was derived from 5-nitro-2-thiophene-carboxaldehyde and 3-iodo-4-methylaniline. 1-(3-iodo-4-methylphenyl)-3-tert-butyl-2H-aminopyrazole (5), prepared from 3-iodo-4-methylaniline.

Solubility
Insoluble in water.

Notes
Light Sensitive .Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

Chemical Identifiers

CAS 35944-64-0
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.05
MDL Number MFCD00047843
InChI Key RRUDMHNAMZFNEK-UHFFFAOYSA-N
Synonym 3-iodo-p-toluidine, benzenamine, 3-iodo-4-methyl, 4-amino-2-iodotoluene, p-toluidine, 3-iodo, 3-iodo-4-methyl-aniline, benzenamine, 3-iodo-4-methyl-9ci, 3-iodo-4-methyl-phenylamine, pubchem3314, 3-iodo-4-methyl aniline, acmc-209ik4
PubChem CID 118889
IUPAC Name 3-iodo-4-methylaniline
SMILES CC1=CC=C(N)C=C1I

Specifications

Melting Point 34°C to 37°C
Quantity 5 g
UN Number UN2811
Beilstein 2353567
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 233.05
Percent Purity 98%
Chemical Name or Material 3-Iodo-4-methylaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 252-803-2
RTECSNumber XU7700000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only