missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodo-4-methoxyaniline 98.0+%, TCI America™
Supplier: TCI America I071625G
Specifications
| 3-Iodo-4-methoxyaniline | |
| 74°C | |
| MFCD06656566 | |
| 4-Amino-2-iodoanisole | |
| COC1=C(I)C=C(N)C=C1 | |
| 249.05 | |
| 249.05 | |
| Crystalline Powder |
| 74587-12-5 | |
| C7H8INO | |
| 25 g | |
| UHPNLGCUIGEZRB-UHFFFAOYSA-N | |
| 3-iodo-4-methoxyaniline | |
| 22054596 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 74587-12-5 | |
| 249.05 | |
| UHPNLGCUIGEZRB-UHFFFAOYSA-N | |
| 22054596 | |
| COC1=C(I)C=C(N)C=C1 |
| C7H8INO | |
| MFCD06656566 | |
| 4-Amino-2-iodoanisole | |
| 3-iodo-4-methoxyaniline |
Safety and Handling
TSCA : No