missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-4-methoxyaniline 98.0+%, TCI America™

Catalog No. I07165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07165G 5 g
I071625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07165G Supplier TCI America Supplier No. I07165G

Chemical Identifiers

CAS 74587-12-5
Molecular Formula C7H8INO
Molecular Weight (g/mol) 249.05
MDL Number MFCD06656566
InChI Key UHPNLGCUIGEZRB-UHFFFAOYSA-N
Synonym 4-Amino-2-iodoanisole
PubChem CID 22054596
IUPAC Name 3-iodo-4-methoxyaniline
SMILES COC1=C(I)C=C(N)C=C1

Specifications

Melting Point 74°C
Quantity 5 g
Formula Weight 249.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodo-4-methoxyaniline