missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-2-propynyl N-Butylcarbamate 97.0+%, TCI America™

$244.19 - $611.41

Chemical Identifiers

CAS 55406-53-6
Molecular Formula C8H12INO2
Molecular Weight (g/mol) 281.093
MDL Number MFCD00072438
InChI Key WYVVKGNFXHOCQV-UHFFFAOYSA-N
Synonym 3-iodoprop-2-yn-1-yl butylcarbamate, iodopropynyl butylcarbamate, iodocarb, ipbc, woodlife, 3-iodo-2-propynyl butylcarbamate, 1-iodoprop-1-yn-3-yl n-n-butylcarbamate, troysan kk-108a, carbamic acid, butyl-, 3-iodo-2-propynyl ester, troysan polyphase anti-mildew
PubChem CID 62097
ChEBI CHEBI:83279
IUPAC Name 3-iodoprop-2-ynyl N-butylcarbamate
SMILES CCCCNC(=O)OCC#CI
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I066625G
View Documents
TCI America
I066625G
25 g
Each for $244.19
Only null left
 
I0666100G
View Documents
TCI America
I0666100G
100 g
Each for $611.41
Only null left
 
Description

Description

Specifications

Chemical Identifiers

55406-53-6
281.093
WYVVKGNFXHOCQV-UHFFFAOYSA-N
62097
3-iodoprop-2-ynyl N-butylcarbamate
C8H12INO2
MFCD00072438
3-iodoprop-2-yn-1-yl butylcarbamate, iodopropynyl butylcarbamate, iodocarb, ipbc, woodlife, 3-iodo-2-propynyl butylcarbamate, 1-iodoprop-1-yn-3-yl n-n-butylcarbamate, troysan kk-108a, carbamic acid, butyl-, 3-iodo-2-propynyl ester, troysan polyphase anti-mildew
CHEBI:83279
CCCCNC(=O)OCC#CI

Specifications

55406-53-6
White
MFCD00072438
2811
WYVVKGNFXHOCQV-UHFFFAOYSA-N
3-iodoprop-2-ynyl N-butylcarbamate
62097
281.09
Crystalline Powder
69°C
C8H12INO2
25 g
3-iodoprop-2-yn-1-yl butylcarbamate, iodopropynyl butylcarbamate, iodocarb, ipbc, woodlife, 3-iodo-2-propynyl butylcarbamate, 1-iodoprop-1-yn-3-yl n-n-butylcarbamate, troysan kk-108a, carbamic acid, butyl-, 3-iodo-2-propynyl ester, troysan polyphase anti-mildew
CCCCNC(=O)OCC#CI
281.093
CHEBI:83279
≥97.0% (GC)
3-Iodo-2-propynyl N-Butylcarbamate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3456

RTECSNumber : EZ0950000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'