missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodo-2-methoxypyridine 98.0+%, TCI America™
$75.08 - $275.30
Chemical Identifiers
| CAS | 112197-15-6 |
|---|---|
| Molecular Formula | C6H6INO |
| Molecular Weight (g/mol) | 235.024 |
| MDL Number | MFCD03094946 |
| InChI Key | BXCHJERCAUZLOE-UHFFFAOYSA-N |
| Synonym | 3-iodo-2-methoxy-pyridine, 2-methoxy-3-iodopyridine, pyridine, 3-iodo-2-methoxy, pubchem6586, acmc-1cuph, ksc490s4l, 3-iodo-2-methoxypyridine |
| PubChem CID | 7009497 |
| IUPAC Name | 3-iodo-2-methoxypyridine |
| SMILES | COC1=C(C=CC=N1)I |
Chemical Identifiers
| 112197-15-6 | |
| 235.024 | |
| BXCHJERCAUZLOE-UHFFFAOYSA-N | |
| 7009497 | |
| COC1=C(C=CC=N1)I |
| C6H6INO | |
| MFCD03094946 | |
| 3-iodo-2-methoxy-pyridine, 2-methoxy-3-iodopyridine, pyridine, 3-iodo-2-methoxy, pubchem6586, acmc-1cuph, ksc490s4l, 3-iodo-2-methoxypyridine | |
| 3-iodo-2-methoxypyridine |
Specifications
| 112197-15-6 | |
| C6H6INO | |
| 1 g | |
| BXCHJERCAUZLOE-UHFFFAOYSA-N | |
| 3-iodo-2-methoxypyridine | |
| 7009497 | |
| ≥98.0% (GC,T) | |
| 3-Iodo-2-methoxypyridine |
| 85°C | |
| MFCD03094946 | |
| 3-iodo-2-methoxy-pyridine, 2-methoxy-3-iodopyridine, pyridine, 3-iodo-2-methoxy, pubchem6586, acmc-1cuph, ksc490s4l, 3-iodo-2-methoxypyridine | |
| COC1=C(C=CC=N1)I | |
| 235.024 | |
| 235.02 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator