accessibility menu, dialog, popup

UserName

H3CONI

CAS RN 112197-15-6

IUPAC Name: 3-iodo-2-methoxypyridine
Synonyms: 3-iodo-2-methoxypyridine
Molecular Weight (g/mol): 235.02
Molecular Formula: C6H6INO
InChi Key: BXCHJERCAUZLOE-UHFFFAOYSA-N
SMILES: COC1=NC=CC=C1I
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results