missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-2-methoxypyridine 98.0+%, TCI America™

Catalog No. I09245G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09245G 5 g
I09241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09245G Supplier TCI America Supplier No. I09245G

Chemical Identifiers

CAS 112197-15-6
Molecular Formula C6H6INO
Molecular Weight (g/mol) 235.024
MDL Number MFCD03094946
InChI Key BXCHJERCAUZLOE-UHFFFAOYSA-N
Synonym 3-iodo-2-methoxy-pyridine, 2-methoxy-3-iodopyridine, pyridine, 3-iodo-2-methoxy, pubchem6586, acmc-1cuph, ksc490s4l, 3-iodo-2-methoxypyridine
PubChem CID 7009497
IUPAC Name 3-iodo-2-methoxypyridine
SMILES COC1=C(C=CC=N1)I

Specifications

Boiling Point 85°C
Quantity 5 g
Formula Weight 235.02
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Iodo-2-methoxypyridine
TSCA No
Recommended Storage Refrigerator