missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodo-2-methoxypyridine 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 85°C |
| Quantity | 1 g |
| Formula Weight | 235.02 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Iodo-2-methoxypyridine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |