missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodo-2,4,5-trimethylthiophene (stabilized with Copper chip) 96.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 87°C |
| Quantity | 1 g |
| Formula Weight | 252.11 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Iodo-2,4,5-trimethylthiophene (stabilized with Copper chip) |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |