accessibility menu, dialog, popup

UserName

CH3SIH3CCH3

CAS RN 60813-84-5

IUPAC Name: 3-iodo-2,4,5-trimethylthiophene
Synonyms: 3-iodo-2,4,5-trimethylthiophene
Molecular Weight (g/mol): 252.11
Molecular Formula: C7H9IS
InChi Key: FYPOBSHIONYIKX-UHFFFAOYSA-N
SMILES: CC1=C(C)C(I)=C(C)S1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results