missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-1H-pyrazole, 97%

Catalog No. AAH3646503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3646503 1 g
AAH3646506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3646503 Supplier Thermo Scientific Chemicals Supplier No. H3646503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4522-35-4 | C3H3IN2 | 193.98 g/mol

3-Iodo-1H-pyrazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodo-1H-pyrazole is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents, acids and bases.

Chemical Identifiers

CAS 4522-35-4
Molecular Formula C3H3IN2
Molecular Weight (g/mol) 193.98
MDL Number MFCD12022324,MFCD01463487
InChI Key RUKDVLFJSMVBLV-UHFFFAOYSA-N
Synonym 3-iodo-1h-pyrazole, 3-iodopyrazole, iodopyrazole, 1h-pyrazole, 3-iodo, 3-iodo-2h-pyrazole, iodo-pyrazole, 5-iodopyrazole, acmc-2097sa
PubChem CID 1239830
IUPAC Name 5-iodo-1H-pyrazole
SMILES IC1=CC=NN1

Specifications

Melting Point 69°C to 72°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 193.98
Percent Purity 97%
Chemical Name or Material 3-Iodo-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only