accessibility menu, dialog, popup

UserName

CAS RN 4522-35-4

IUPAC Name: 5-iodo-1H-pyrazole
Synonyms: 3-iodo-2H-pyrazole
Molecular Weight (g/mol): 193.98
Molecular Formula: C3H3IN2
InChi Key: RUKDVLFJSMVBLV-UHFFFAOYSA-N
SMILES: IC1=CC=NN1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)