Learn More
3-Indoxyl sulfate potassium salt, 97%
CAS: 2642-37-7 | C8H6KNO4S | 251.297 g/mol
Supplier: Thermo Scientific Chemicals A1707903
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Indoxyl sulfate potassium salt | |
| 165°C (decomposition) | |
| MFCD00037931 | |
| 3921324 | |
| potassium 1h-indol-3-yl sulfate, urinary indican, potassium indol-3-yl sulfate, indican urinary, indol-3-yl potassium sulfate, indoxyl sulfate potassium salt, potassium indol-3-yl sulphate, unii-567hmw942w, indol-3-ol, potassium sulfate, indol-3-yl sulfate, potassium salt | |
| C1=CC=C2C(=C1)C(=CN2)OS(=O)(=O)[O-].[K+] | |
| 251.297 | |
| 251.31 |
| 2642-37-7 | |
| C8H6KNO4S | |
| 1 g | |
| Light sensitive | |
| MDAWATNFDJIBBD-UHFFFAOYSA-M | |
| potassium;1H-indol-3-yl sulfate | |
| 5177095 | |
| 97% |
Chemical Identifiers
| 2642-37-7 | |
| 251.297 | |
| MDAWATNFDJIBBD-UHFFFAOYSA-M | |
| 5177095 | |
| C1=CC=C2C(=C1)C(=CN2)OS(=O)(=O)[O-].[K+] |
| C8H6KNO4S | |
| MFCD00037931 | |
| potassium 1h-indol-3-yl sulfate, urinary indican, potassium indol-3-yl sulfate, indican urinary, indol-3-yl potassium sulfate, indoxyl sulfate potassium salt, potassium indol-3-yl sulphate, unii-567hmw942w, indol-3-ol, potassium sulfate, indol-3-yl sulfate, potassium salt | |
| potassium;1H-indol-3-yl sulfate |
Safety and Handling
EINECSNumber : 220-145-5
RTECSNumber : NM3200000
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only