accessibility menu, dialog, popup

UserName

CAS RN 2642-37-7

IUPAC Name: potassium 1H-indol-3-yl sulfate
Synonyms: potassium indol-3-yl sulfate
Molecular Weight (g/mol): 251.3
Molecular Formula: C8H6KNO4S
InChi Key: MDAWATNFDJIBBD-UHFFFAOYSA-M
SMILES: [K+].[O-]S(=O)(=O)OC1=CNC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results