missing translation for 'onlineSavingsMsg'
Learn More

3-Indoxyl sulfate potassium salt, 97%

Catalog No. AAA1707901
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1707901 0.25 g
AAA1707903 1 g
AAA1707906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1707901 Supplier Thermo Scientific Chemicals Supplier No. A1707901
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2642-37-7 | C8H6KNO4S | 251.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2642-37-7
Molecular Formula C8H6KNO4S
Molecular Weight (g/mol) 251.297
MDL Number MFCD00037931
InChI Key MDAWATNFDJIBBD-UHFFFAOYSA-M
Synonym potassium 1h-indol-3-yl sulfate, urinary indican, potassium indol-3-yl sulfate, indican urinary, indol-3-yl potassium sulfate, indoxyl sulfate potassium salt, potassium indol-3-yl sulphate, unii-567hmw942w, indol-3-ol, potassium sulfate, indol-3-yl sulfate, potassium salt
PubChem CID 5177095
IUPAC Name potassium;1H-indol-3-yl sulfate
SMILES C1=CC=C2C(=C1)C(=CN2)OS(=O)(=O)[O-].[K+]

Specifications

Melting Point 165°C (decomposition)
Quantity 0.25 g
Beilstein 3921324
Sensitivity Light sensitive
Formula Weight 251.31
Percent Purity 97%
Chemical Name or Material 3-Indoxyl sulfate potassium salt
EINECSNumber 220-145-5
RTECSNumber NM3200000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only