missing translation for 'onlineSavingsMsg'
Learn More

3-Indolemethanol 98.0+%, TCI America™

Catalog No. p-7131441
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04965G 5 g
I049625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04965G Supplier TCI America Supplier No. I04965G
Only null left

Chemical Identifiers

CAS 700-06-1
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00005632
InChI Key IVYPNXXAYMYVSP-UHFFFAOYSA-N
Synonym indole-3-carbinol, 3-indolemethanol, 1h-indol-3-yl methanol, indole-3-methanol, 1h-indole-3-methanol, 3-hydroxymethylindole, 3-indolylcarbinol, indinol, 3-indole methanol, indole 3 carbinol
PubChem CID 3712
ChEBI CHEBI:24814
IUPAC Name 1H-indol-3-ylmethanol
SMILES C1=CC=C2C(=C1)C(=CN2)CO

Specifications

Quantity 5 g
Formula Weight 147.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Indolemethanol
RTECSNumber NL9483000
TSCA No
Recommended Storage Freezer