accessibility menu, dialog, popup

UserName

HOHN

CAS RN 700-06-1

IUPAC Name: (1H-indol-3-yl)methanol
Synonyms: indole-3-carbinol indinol I3C cpd
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: IVYPNXXAYMYVSP-UHFFFAOYSA-N
SMILES: OCC1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)