missing translation for 'onlineSavingsMsg'
Learn More

3-Indolemethanol, 97%

Catalog No. aal1576206
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AAL1576206 5 g
AAL1576203 1 g
2 options

Catalog No. AAL1576206

Supplier: Thermo Scientific Chemicals L1576206

Only null left

CAS: 700-06-1 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 700-06-1
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00005632
InChI Key IVYPNXXAYMYVSP-UHFFFAOYSA-N
Synonym indole-3-carbinol, 3-indolemethanol, 1h-indol-3-yl methanol, indole-3-methanol, 1h-indole-3-methanol, 3-hydroxymethylindole, 3-indolylcarbinol, indinol, 3-indole methanol, indole 3 carbinol
PubChem CID 3712
ChEBI CHEBI:24814
IUPAC Name 1H-indol-3-ylmethanol
SMILES C1=CC=C2C(=C1)C(=CN2)CO

Specifications

Melting Point 96°C to 99°C
Quantity 5 g
Beilstein 121323
Formula Weight 147.18
Percent Purity 97%
Chemical Name or Material 3-Indolemethanol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-836-2
RTECSNumber NL9483000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only