missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Indoleacrylic Acid 98.0+%, TCI America™
$123.03 - $363.49
Chemical Identifiers
| CAS | 1204-06-4 |
|---|---|
| Molecular Formula | C11H9NO2 |
| Molecular Weight (g/mol) | 187.198 |
| MDL Number | MFCD00005633 |
| InChI Key | PLVPPLCLBIEYEA-AATRIKPKSA-N |
| Synonym | 3-indoleacrylic acid, trans-3-indoleacrylic acid, indole-3-acrylic acid, 3-indolylacrylic acid, indole-3beta-acrylic acid, 3-1h-indol-3-yl acrylic acid, indole-3-crylic acid, 2e-3-1h-indol-3-yl prop-2-enoic acid, trans-3-1h-indol-3-yl acrylic acid, indoleacrylic acid van |
| PubChem CID | 5375048 |
| IUPAC Name | (E)-3-(1H-indol-3-yl)prop-2-enoic acid |
| SMILES | C1=CC=C2C(=C1)C(=CN2)C=CC(=O)O |
Chemical Identifiers
| 1204-06-4 | |
| 187.198 | |
| PLVPPLCLBIEYEA-AATRIKPKSA-N | |
| 5375048 | |
| C1=CC=C2C(=C1)C(=CN2)C=CC(=O)O |
| C11H9NO2 | |
| MFCD00005633 | |
| 3-indoleacrylic acid, trans-3-indoleacrylic acid, indole-3-acrylic acid, 3-indolylacrylic acid, indole-3beta-acrylic acid, 3-1h-indol-3-yl acrylic acid, indole-3-crylic acid, 2e-3-1h-indol-3-yl prop-2-enoic acid, trans-3-1h-indol-3-yl acrylic acid, indoleacrylic acid van | |
| (E)-3-(1H-indol-3-yl)prop-2-enoic acid |
Specifications
| 1204-06-4 | |
| MFCD00005633 | |
| 3-indoleacrylic acid, trans-3-indoleacrylic acid, indole-3-acrylic acid, 3-indolylacrylic acid, indole-3beta-acrylic acid, 3-1h-indol-3-yl acrylic acid, indole-3-crylic acid, 2e-3-1h-indol-3-yl prop-2-enoic acid, trans-3-1h-indol-3-yl acrylic acid, indoleacrylic acid van | |
| C1=CC=C2C(=C1)C(=CN2)C=CC(=O)O | |
| 187.198 | |
| 187.20 | |
| Crystalline Powder |
| C11H9NO2 | |
| 1 g | |
| PLVPPLCLBIEYEA-AATRIKPKSA-N | |
| (E)-3-(1H-indol-3-yl)prop-2-enoic acid | |
| 5375048 | |
| ≥98.0% (HPLC,T) | |
| 3-Indoleacrylic Acid |
Safety and Handling
RTECSNumber : NL3680000
TSCA : Yes