accessibility menu, dialog, popup

UserName

CAS RN 1204-06-4

HOOHN(Z)

CAS RN 1204-06-4

IUPAC Name: (2Z)-3-(1H-indol-3-yl)prop-2-enoic acid
Synonyms: (2Z)-3-(1H-indol-3-yl)prop-2-enoic acid
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: PLVPPLCLBIEYEA-WAYWQWQTSA-N
SMILES: OC(=O)\C=C/C1=CNC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results