missing translation for 'onlineSavingsMsg'
Learn More

3-Indoleacetic Acid 98.0+%, TCI America™

Catalog No. p-7131160
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
I00225G 5 g
I002225G 25 g
2 options

Catalog No. I00225G

Supplier: TCI America I00225G

Only null left

Chemical Identifiers

CAS 87-51-4
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00005636
InChI Key SEOVTRFCIGRIMH-UHFFFAOYSA-N
Synonym indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon a
PubChem CID 802
ChEBI CHEBI:16411
IUPAC Name 2-(1H-indol-3-yl)acetic acid
SMILES OC(=O)CC1=CNC2=CC=CC=C12

Specifications

Melting Point 168°C
Quantity 5 g
Formula Weight 175.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Indoleacetic Acid
EINECSNumber (5)-6618
RTECSNumber NL3150000
TSCA Yes