missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxytyrosol, ≥98.0% (GC), MilliporeSigma™ Supelco™
Description
Food and Beverages
Specifications
Specifications
| CAS | 10597-60-1 |
| Linear Formula | C8H10O3 |
| MDL Number | MFCD01320529 |
| Synonym | 2-(3,4-Dihydroxyphenyl)ethanol; 3,4-Dihydroxyphenethyl alcohol; DOPET; Homoprotocatechuyl alcohol |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 154.16 |
| Percent Purity | ≥98.0% (GC) |
| Grade | Analytical Standard |