missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypyridine 98.0+%, TCI America™

Catalog No. p-7130490
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H033125G 25 g
H0331100G 100 g
H0331500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H033125G Supplier TCI America Supplier No. H033125G
Only null left

Chemical Identifiers

CAS 109-00-2
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.101
MDL Number MFCD00006378
InChI Key GRFNBEZIAWKNCO-UHFFFAOYSA-N
Synonym 3-hydroxypyridine, 3-pyridinol, 3-pyridol, 3-pyridone, 3-oxopyridine, beta-hydroxypyridine, m-hydroxypyridine, 3-pyridyl alcohol, 3-hydoxypyridine, unii-4kbe4p5b6s
PubChem CID 7971
IUPAC Name pyridin-3-ol
SMILES C1=CC(=CN=C1)O

Specifications

Melting Point 126°C
Boiling Point 180°C
Quantity 25 g
Formula Weight 95.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxypyridine
EINECSNumber (5)-0722
RTECSNumber UU7701400
TSCA Yes