accessibility menu, dialog, popup

UserName

OHN

CAS RN 109-00-2

IUPAC Name: pyridin-3-ol
Synonyms: 2-pyridone
Molecular Weight (g/mol): 95.1
Molecular Formula: C5H5NO
InChi Key: GRFNBEZIAWKNCO-UHFFFAOYSA-N
SMILES: OC1=CC=CN=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (3)
  • (5)