missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypyridine, 98%

Catalog No. p-7021537
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Catalog No. Quantity
AAA1391018 50 g
AAA1391022 100 g
AAA1391036 500 g
3 options

Catalog No. AAA1391018

Supplier: Thermo Scientific Chemicals A1391018

Only null left

CAS: 109-00-2 | C5H5NO | 95.101 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-00-2
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.101
MDL Number MFCD00006378
InChI Key GRFNBEZIAWKNCO-UHFFFAOYSA-N
Synonym 3-hydroxypyridine, 3-pyridinol, 3-pyridol, 3-pyridone, 3-oxopyridine, beta-hydroxypyridine, m-hydroxypyridine, 3-pyridyl alcohol, 3-hydoxypyridine, unii-4kbe4p5b6s
PubChem CID 7971
IUPAC Name pyridin-3-ol
SMILES C1=CC(=CN=C1)O

Specifications

Melting Point 125°C to 128°C
Boiling Point 180°C (22 mmHg)
Flash Point 173°C (343°F)
Quantity 50 g
Beilstein 105699
Sensitivity Light sensitive
Formula Weight 95.1
Percent Purity 98%
Chemical Name or Material 3-Hydroxypyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 203-637-4
RTECSNumber UU7701400
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only